BRAND® Dispensette® S Trace Analysis Bottletop Dispensers, BrandTech®
Supplier: Brandtech
4640040 4640240 4640041 4640241
10019-026EA 2935.8 CAD
10019-026 10019-028 10019-030 10019-032
BRAND® Dispensette® S Trace Analysis Bottletop Dispensers, BrandTech®
Bottle Top Dispensers
The new Dispensette® S Trace Analysis features many of the same safety and convenience features of the Dispensette® S, but is optimized for the needs of dispensing high purity reagents.
  • Optimized for high purity reagents
  • Wiping seal piston
  • Minimal reagent exposure
Designed to attach directly to a reagent bottle to eliminate poured transfer, the Dispensette® S Trace Analysis features a wiping seal piston to minimize reagent exposure
Instruments feature a sliding pointer, well suited for rapid volume changes from one end of the range to the other. AMore Product Information