Formvar Carbon Support Film on Thin Bar Hexagonal Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
FCFTH400-CU-EB FCFTH400-CU-EC FCFTH300-CU-SB FCFTH300-CU-SC FCFTH300-CU-UA FCFTH300-CU-UB FCFTH300-CU-UC FCFTH400-CU-SB FCFTH400-CU-SC FCFTH400-CU-UA FCFTH400-CU-UB FCFTH400-CU-UC FCFTH300-CU-TA FCFTH300-CU-TB FCFTH300-CU-TC FCFTH400-CU-TA FCFTH400-CU-TB FCFTH400-CU-TC FCFTH200-CU-SB FCFTH200-CU-SC FCFTH200-CU-UA FCFTH200-CU-UB FCFTH200-CU-UC FCFTH300-CU-EA FCFTH300-CU-EB FCFTH300-CU-EC FCFTH400-CU-EA FCFTH200-CU-TA FCFTH200-CU-TB FCFTH200-CU-TC FCFTH200-CU-EA FCFTH200-CU-EB FCFTH200-CU-EC FCFTH200-NI-UC FCFTH300-NI-SB FCFTH300-NI-SC FCFTH300-NI-UA FCFTH300-NI-UB FCFTH300-NI-UC FCFTH400-NI-SB FCFTH400-NI-SC FCFTH400-NI-UA FCFTH400-NI-UB FCFTH400-NI-UC FCFTH600-NI-UA FCFTH200-NI-TA FCFTH200-NI-TB FCFTH200-NI-TC FCFTH300-NI-TA FCFTH300-NI-TB FCFTH300-NI-TC FCFTH400-NI-TA FCFTH400-NI-TB FCFTH400-NI-TC FCFTH200-NI-EA FCFTH200-NI-EB FCFTH200-NI-EC FCFTH300-NI-EA FCFTH300-NI-EB FCFTH300-NI-EC FCFTH400-NI-EA FCFTH200-AU-SB FCFTH300-AU-SB FCFTH400-AU-SB FCFTH200-NI-SB FCFTH200-NI-SC FCFTH200-NI-UA FCFTH200-NI-UB FCFTH400-NI-EB FCFTH400-NI-EC
103303-962EA 284.21 CAD
103303-962 103303-964 103304-314 103304-316 103304-318 103304-320 103304-322 103304-324 103304-326 103304-328 103304-330 103304-332 103304-514 103304-564 103304-566 103304-568 103304-570 103304-572 103304-728 103304-730 103304-732 103304-734 103304-736 103304-738 103304-740 103304-742 103304-744 103304-964 103304-966 103304-968 103305-020 103305-022 103305-024 103305-170 103305-172 103305-174 103305-176 103305-178 103305-180 103305-182 103305-184 103305-186 103305-188 103305-190 103305-196 103305-252 103305-254 103305-256 103305-258 103305-260 103305-262 103305-264 103305-266 103305-268 103305-332 103305-334 103305-336 103305-338 103305-340 103305-342 103305-344 103305-652 103305-662 103305-672 103305-880 103305-882 103305-884 103305-886 103305-936 103305-938
Formvar Carbon Support Film on Thin Bar Hexagonal Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
The formvar coated grids are stabilized with an evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra-thin sections.
  • Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
  • Carbon coatings thickness of 1 nm, 3 to 4 nm, and 20 to 30 nm
  • Select copper, nickel, or gold grids in 200, 300, 400, or 600 mesh
  • Optically checked followed by batch testing in the EM
All films are laid on the shiny side of the grid.
Packaging: Packed in grid storage box.