Avantor Sterile Polycarbonate Single-Use Bottle Assemblies, Avantor Fluid Handling
Supplier: VWR International
10830-302 10830-304 10830-306 10830-696 10830-698 75835-990
10830-302CS 4868.45 CAD
10830-302 10830-304 10830-306 10830-696 10830-698 75835-990
Avantor Sterile Polycarbonate Single-Use Bottle Assemblies, Avantor Fluid Handling
Bottles Cell Culture Bottles
Sterile polycarbonate containers with weldable tubing provide the maximum amount of flexibility and reliability.
  • Sterile bottles come individually bagged and process ready
  • Multiple bottle sizes allow for process-specific flexibility
  • Gamma irradiated to Sterility Assurance Level (SAL) 10⁻⁶
These vessels are assembled in an ISO Class 7 cleanroom and are manufactured from USP Class VI materials.
When used with sterile Avantor tube sets, custom possibilities are endless.
Assemblies have been designed with a broad spectrum of applications in mind,More Product Information