Accessories for Q-sep® 3000 Centrifuge, Restek
Supplier: Restek
26234 26249
10015-330PK 195.92 CAD
10015-330 10015-332 10015-334 10015-446
Accessories for Q-sep® 3000 Centrifuge, Restek
Centrifuges Benchtop Centrifuges
Accessories include 50 mL tube carrier, 50 mL conical tube insert, 15 mL 4-place tube carrier (four 15 mL tubes), and 2 mL tube adapters.
  • Improve functionality of Q-sep® 3000 Centrifuge
  • Durable construction of carriers and inserts ensures long lifetime