Sample Cylinders, Ultra-High Pressure (Stainless Steel & Sulfinert®-Treated), Restek
Supplier: Restek
22111 22111-PI 22112 22112-PI 22113 22113-PI 22929-PI 22927 22927-PI 22928 22928-PI 22929
10145-726EA 2036.14 CAD
10145-726 10145-728 10145-730 10145-732 10145-734 10145-736 10145-740 10145-834 10145-836 10145-838 10145-840 10145-842
Sample Cylinders, Ultra-High Pressure (Stainless Steel & Sulfinert®-Treated), Restek
Cylinders Hydrometer Cylinders
Swagelok® sample cylinders are made of 304L and 316L stainless steel to resist corrosion and DOT rated to 1,800 and 5,000 psig (TPED cylinders rated to 1,450 and 4,350 psig), which allows sampling at gas wellheads as well as on-site refineries.
  • 316L stainless steel; DOT rating to 5,000 psig (TPED cylinders to 4,350 psig)
  • Range of cylinder sizes, 150 cc to 500 cc
  • All cylinders have 1/4" female NPT threads on both ends
  • TPED compliant cylinders available for EU community
Each cylinder is hydrostatically tested to at least 5/3 the working pressure.