Q-sep® QuEChERS Extraction Salts, Restek
Supplier: Restek
25851 25847 25848 25850 25849 25852 25846
76204-758PK 447.76 CAD
76204-758 76204-766 76204-768 76204-762 76204-764 76204-760 76204-770
Q-sep® QuEChERS Extraction Salts, Restek
Protein Extraction Protein Extraction Reagents
QuEChERS methods are fast, easy, and cost-effective, and Restek Q-sep® products make QuEChERS procedures even simpler.
  • Free-flowing salts transfer easily and completely
  • Easy-open packets eliminate the need for a second empty tube for salt transfer
  • Convenient slim packets fit perfectly into tubes to prevent spills
  • Ready-to-use tubes; no glassware required
  • Pre-weighed, ultra-pure extraction salts
  • Ideal for original unbuffered, AOAC (2007.01), and European (EN 15662) QuEChERS methods
No specialized glassware is required when using Q-sep® extraction packets and tubes. Free-flowing extraction salts and salt packets fit easily into the extraction tubes to make transferring salts to sample mess-free and easy.