Extraction Cell Caps and Replacement Parts for ASE® Systems, Restek
Supplier: Restek
26188 26096 26180 26134 26187 26174 26184 25997 26170 26169 26167 25959 26166 26100
10015-278PK 533.76 CAD
10015-278 10015-264 10015-298 10015-276 10015-314 10015-378 10015-422 10015-292 10015-284 10015-296 10015-392 10015-318 10015-418 10015-316
Extraction Cell Caps and Replacement Parts for ASE® Systems, Restek
Chromatography Supplies Chromatography Spare Parts
Accelerated solvent extraction is a common technique for fast and reliable extraction of organic materials from solid matrices, using EPA SW-846 Method 3545, Pressurized Fluid Extraction (PFE).
  • Smooth inner surfaces for easier cleaning
  • Caps include frit, PEEK washer, PTFE O-ring, threaded insert, and snap ring
Restek offers a wide range of replacement parts to keep your extraction system running smoothly. All parts are economically priced to save you money and are designed to meet or exceed the performance of the original manufacturer’s parts.