Q-sep® QuEChERS dSPE Tubes for Extract Cleanup, Restek
Supplier: Restek
26224 26222 26223 26225 26226 26244 26215 26216 26217 26218 26219 26243 26220 26221 26242 26245
89207-066PK 337.8 CAD
89207-066 10057-972 10057-974 10057-976 10057-978 10057-984 10059-524 10059-526 10059-528 10059-530 10059-532 10803-400 10015-310 10015-320 10015-340 10015-342
Q-sep® QuEChERS dSPE Tubes for Extract Cleanup, Restek
Tubes Gas Sampling Tubes
QuEChERS methods are fast, easy, and cost-effective, and Restek Q-sep® products make QuEChERS procedures even simpler.
  • Packaged in foil subpacks of 10 for enhanced protection and storage stability
  • Multiple sorbents are used to extract different types of interferences
  • MgSO₄ removes excess water
  • Ready-to-use tubes, no glassware required
  • Preweighed, ultra-pure sorbents
  • Support original unbuffered, AOAC (2007.01), European (EN 15662), and mini-multiresidue QuEChERS methods
  • C18 - removes non polar interferences
All extraction salts, sorbents, and sample tubes are included - no specialized equipment or glassware is required.
PSA - primary and secondary amine; removes sugars, fatty acids, organic acids, and anthocyanine pigments
GCB - graphitized carbon black; removes pigments, sterols, and non polar interferences
Prepare samples more efficiently with a complete line of QuEChERS supplies from Restek.