Sample Collection Bottles for ASE® 100/300 Systems, 250 ml, Restek
Supplier: Restek
26191 26260
10015-304PK 73.03 CAD
10015-304 10015-450
Sample Collection Bottles for ASE® 100/300 Systems, 250 ml, Restek
Bottles Glass Bottles
Cleaned, ready-to-use sample collection bottles are suitable for use with ASE® 100/300 systems.
  • Cleaned, assembled, and ready to use
  • Collection bottles with volume of 250 mL
  • Clear or amber glass
  • Caps and PTFE-lined septa included
For use with ASE® 100/300 systems.