Resprep® Tools for ASE® Systems, Restek
Supplier: Restek
26181
10015-286PK 365.47 CAD
10015-286
Resprep® Tools for ASE® Systems, Restek
Extractors Automatic Extractor Systems
Resprep® tools are specialized to simplify routine chores and suitable for ASE 100/200/300.
  • Use to insert filter in extraction cell or O-ring in cell cap
  • Fits all extraction cells, except 1 mL size
Handle is easy to grip, and various attachments alllow for compatibility with extraction cells of all sizes.
Delivery information: Includes Resprep tool handle and filter insertion attachments.