Gas Sample Loops (Sulfinert®-Treated), Restek
Supplier: Restek
22841 22842 22844 22845 22846 22847 22848 22849 22850 22843
10058-804EA 658.54 CAD
10058-804 10058-806 10058-808 10058-810 10058-812 10058-814 10058-816 10058-818 10058-820 10803-302
Gas Sample Loops (Sulfinert®-Treated), Restek
Cylinders Hydrometer Cylinders
Sulfinert® treatment eliminates active sites in the valve or loop for better recovery of active compounds.
  • Ideal for samples containing low concentrations of sulfur or other active compounds
  • Sample loop sizes from 5 µL to 5 mL
These gas sample loops have fittings sized at 1/16" with an internal diameter of 1 mm, for use with “W Type” valves.