Method Specific SPE Cartridges, Restek
Supplier: Restek
26032 24049 25499 26093
10803-386PK 457.99 CAD
10803-386 10015-362 10015-372 10015-416
Method Specific SPE Cartridges, Restek
Chromatography Columns
These cartridges have been specifically designed to provide consistent and reproducible results for the method or application stated.
  • Method compatibility differs between models
  • Highly consistent and reproducible results
  • Application-specific cartridges
10015-416: Extraction of explosive compounds (similar to EPA method 8095 and 8330 list) from water samples.
10015-404: Separation of aliphatic and aromatic hydrocarbons into distinct extract fractions. Specially treated to reduce contaminants and increase capacity. Silica packing.
13501-554: Extraction of endothall from aqueous samples. Weak anion exchange resin (BioRex 5) packing.
10803-386: For use in EPA method 521: Nitrosamines in Drinking Water and EPA method 522 for 1,4-Dioxane in Drinking Water. Activated charcoal packing.
10015-362: High-capacity cleanup of butyl and phenyl tin compounds from soil, water, and biota. Mixed bed.