Science Take-Out® Atoms, Isotopes And Ions
Supplier: SCIENCE TAKE-OUT, LLC
STO-142 STO-142U STO-142-NOTG
470157-258EA 27.2 CAD
470157-258 470157-260 470157-259
Science Take-Out® Atoms, Isotopes And Ions
Educational Materials Chemistry Educational Materials Ionic Properties Learning Activities
Use Colored Chips To Model Atomic Structure.
  • Available As A Single Kit With Materials For 1 Student Or Group
  • Unassembled 10 Pack For Additional Savings
  • No Additional Equipment Needed
Use colored chips and the information from a periodic table to model the sub-atomic particles (protons, neutrons, and electrons) in atoms, isotopes, and ions. Model the electron configuration within the electron energy levels. Learn how changing the number of protons, neutrons, or electrons leads to different elements, isotopes, or ions. This complete "dry lab" activityMore Product Information