Dieffenbach Bulldog Clamp, OR Grade, Sklar
Supplier: Sklar
17-3815 17-3817 17-3915 17-3917
10192-450EA 166.36 CAD
10192-450 10194-808 10194-810 10194-812
Dieffenbach Bulldog Clamp, OR Grade, Sklar
Clamps General Purpose Clamps
Sklar's Dieffenbach Bulldog Clamps are small, bayonet-style forceps most commonly used for clamping small vessels
They are often used in cardiovascular procedures. They have finely serrated jaws with rounded tips, that may be slightly curved or straight. A bulldog clamp may also be known as a serrefine.