Mayo Trays, Sklar®
Supplier: Sklar
10-1690 10-1690-12 10-1690-6 10-1691-12 10-1691-6 10-1692 10-1692-12 10-1692-6 10-1693 10-1693-12 10-1693-6 10-1694-12 10-1694-6 10-1695 10-1695-12 10-1695-6
10193-192EA 270.24 CAD
10193-192 10193-194 10193-196 10193-198 10193-200 10193-202 10193-204 10193-206 10193-208 10193-210 10193-212 10193-214 10193-216 10193-218 10193-220 10193-222
Mayo Trays, Sklar®
Trays/Dishes/Pots Instrument Trays/Dishes
The Mayo Tray holds instruments to be used in a surgical procedure
It is typically organized and monitored by the scrub person or surgical technologist assisting in the procedure. It is used in conjunction with a stand so that the tray and instruments are situated above the waist and easily accessible during the procedure. The tray holds instruments that are the most frequently used during any given procedure. The tray is available perforated or non perforated and in a variety of sizes. Stainless steel material.