Orion Dermatoscope, OR Grade, Sklar
Supplier: Sklar
06-6250
10194-704EA 1336.32 CAD
10194-704
Orion Dermatoscope, OR Grade, Sklar
Clinical Diagnostic Supplies Clinical Instruments
Sklar's Orion Dermatoscope is a dermal diagnostic instrument used in the examination of skin lesions
  • Stainless Steel/Plastic
It is an oil immersion/non-polarized instrument that utilizes 10X magnification and illumination to closely examine skin lesions, melanoma, and infections. Oil or petroleum jelly may be used with the graduated plate to provide additional contrast and definition. The handle of the Orion Dermatoscope features rheostats that adjust the light intensity with a simple rotation.