Richardson Retractor, OR Grade, Sklar
Supplier: Sklar
60-1686 60-1691 60-1676 60-1681
10194-248EA 432.89 CAD
10194-248 10197-274 10197-718 10197-720
Richardson Retractor, OR Grade, Sklar
Retractors
Sklar's Richardson Retractor is a hand-held, single-end, right angle retractor used for holding back multiple layers of deep tissue
  • Made of stainless steel
Censitrac Ready products are industry standard.
This retractor is often used in appendectomy procedures, as well as cesarean sections and laparotomy. The blade is right angled, concave, and has a slight lip at the tip. The handle is hollow and features finger ridges for improved grip. The Richardson retractor is often mistaken for a Kelly retractor, however the Richardson retractor has a smaller blade.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.