Micro Trays, Sklar®
Supplier: Sklar
10-1702 10-1716 10-1709
10193-230EA 1734.6 CAD
10193-230 10193-236 10193-238 10193-248 76191-446
Micro Trays, Sklar®
Trays/Dishes/Pots Instrument Trays/Dishes
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Micro Sterilizing Trays are used to hold smaller instruments securely during a sterilization process. Rows of separating pins and silicone hold down bars for locking instruments in place. The tray also has a hinged lid, locking latch, and outside handles.
Order Now
SPECIFICATIONS
Environmentally Preferable