Sera-Mag Streptavidin-Coated Magnetic Beads and SpeedBeads, Cytiva
Supplier: Cytiva
301521310150 301521310350 301521311150 301521410150 301521410350 30152104011150 301521510150 301521510350 301521511150 66152110150 66152110350 66152111150
CA10146-030EA 1752.49 CAD
CA10146-030 CA10146-032 CA10146-034 CA10146-036 CA10146-038 CA10146-040 CA10146-048 CA10146-050 CA10146-052 CA10146-078 CA10146-080 CA10146-082
Sera-Mag Streptavidin-Coated Magnetic Beads and SpeedBeads, Cytiva
Beads Magnetic Beads
Sera-Mag Streptavidin-Coated Magnetic Beads and SpeedBeads exhibit high affinity and sensitivity for biotinylated target molecules, along with fast reaction kinetics, increasing throughput and precision for genomic and proteomic applications.
  • Available with low (2500 to 3500 pmol/mg), medium (3500 to 4500 pmol/mg) or high (4500 to 5500 pmol/mg) nominal biotin binding capacities for optimizing assay development
  • Low dissociation constant to tightly bound ligands
  • Low non-specific binding for greater precision and accuracy
  • Available in Sera-Mag SpeedBeads and
More Product Information