Emasculatome with Cord Stop, OR Grade, Sklar
Supplier: Sklar
93-8109 93-8112 93-8114
10185-500EA 290.94 CAD
10185-500 10185-502 10185-504
Emasculatome with Cord Stop, OR Grade, Sklar
Cutters
Emasculatome with Cord Stop, veterinary instrument used for castrating primarily lambs and goats.
The clamping tips crush the spermatic cords but do not cut the skin or cause damage to internal vessels.