Kelly Retractor with SklarGrip™ Handle, OR Grade, Sklar
Supplier: Sklar
60-1656 60-1661 60-1666 60-1671
10197-710EA 532.73 CAD
10197-710 10197-712 10197-714 10197-716
Kelly Retractor with SklarGrip™ Handle, OR Grade, Sklar
Retractors
Sklar Kelly Retractors are available in two distinct styles
It is available with various blade lengths and can therefore retract several layers of tissue if necessary. This retractor is often confused with the Richardson retractor, however, the Kelly retractor features longer blades. Sklar offers this Kelly retractor with a grip handle that is hollow and features finger ridges and a slight hook at the end for a secure grip and improved comfort.
This Kelly Retractor is a manual, right angle, general retractor used for wound exposure.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.