TC Hegar Vascular Needle Holder, OR Grade, Sklar
Supplier: Sklar
20-4178 20-4175
10185-274EA 557.02 CAD
10185-274 10195-176
TC Hegar Vascular Needle Holder, OR Grade, Sklar
Forceps
Sklar's® TC Hegar finger ring needle holders are used in cardiovascular surgeries for suturing.
Tungsten carbide instruments are designed to stay sharp for a significantly longer period of time than other standard instruments
TC Hegar and Censitrac Ready products are industry standard and constructed of stainless steel.
Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.