Alexander Mastoid Gouge, OR Grade, Sklar
Supplier: Sklar
67-4504 67-4506 67-4508 67-4510 67-4512 67-4514
10198-366EA 311.26 CAD
10198-366 10198-368 10198-370 10198-372 10198-374 10198-376
Alexander Mastoid Gouge, OR Grade, Sklar
Gouges
The Sklar® Alexander Mastoid Gouge is an ENT instrument used to carve away portions of the mastoid bone, typically due to infection.
  • Reusable, Latex-free
  • Non sterile
The gouge has one solid end that is the handle and is tapped on the end with a hammer to push the opposite blade end along and into the bone section to be removed.
The long, scoop-style blade begins at the mid point of the instrument. As the steel thins to create the scoop the thin area also widens to a sharp end that scrapes away the bone.