VWR® TraceableLIVE® Wi-Fi Data Logging Refrigerator / Freezer Thermometers with Remote Notification
Supplier: VWR International
10816-260 10816-262 10816-264 10816-266
10816-260EA 717.95 CAD
10816-260 10816-262 10816-264 10816-266
VWR® TraceableLIVE® Wi-Fi Data Logging Refrigerator / Freezer Thermometers with Remote Notification
Data Loggers
The simple, efficient, and reliable way to stay connected to your critical environments, wherever you go.
  • Monitor temperature and more locally or remotely
  • Alerts: temperature alarm, connectivity interruption, low battery
  • Cloud-based data interface
  • Continuous data transmission and monitoring using SSL encrypted SECURE connection
  • Receive MOBILE PUSH / E-MAIL / TEXT notifications
  • Multiple point temperature calibration—traceable to NIST
  • Unlimited scalability-use TraceableLIVE® in individual labs or an entire network
  • of facilities
Bottle probe: Temperature-buffered glycol bottle probe mimics solution-bottle and solution insulate sensor from rapid changes. Solution mimics temperature of stored samples. Probe is sealed in a miniature bottle (1×2⁻¹/₂") filled with nontoxic glycol. Solution is GRAS (Generally recognized as safe) by the FDA (Food and drug administration).
Bullet probe: Small sized plastic probe with cable 0.19" Ø×0.75" long. Designed for use in air and liquids, the sensor and cable may be completely immersed.
Stainless-steel probe: Fast-response Stainless steel 316 probe displays the slightest temperature change instantly. Readings are updated three times a second. Accurate readings with tip penetration of 1". Probe has a diameter of 0.12", stem length of 6.25", an overall length of 9" and a cable length of 10'.
Audible and visual alarms helps those present in the lab identify any out-of-range conditions. Ideal for use in healthcare and vaccine monitoring (VFC compliant). Works with any refrigerator or freezer. High-impact, chemical-resistant ABS plastic case.
With TraceableLIVE®, stay connected to your lab with your smartphone, tablet, or personal computer. By simply using the TraceableLIVE® app, or visiting Traceablelive.com on any browser: view current environmental conditions, control alarm parameters, view datalogging history, view and generate reports. TraceableLIVE® Standard and Premium subscription plans are now available on website (https://us.vwr.com/store/product/35955573/traceablelive-subscription-coupon-codes-cole-parmer).
Certifications: CE marked. RoHS compliant. Traceable to NIST for accuracy. Multi-point calibration on an individually-numbered Traceable® certificate which assures accuracy from an ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM mutual recognition agreement.
Ordering information: Requires TraceableLIVE subscription. Units are supplied with 2 probes, quick start guide, batteries, wall mount, Velcro®, magnetic strips and an ISO 17025 Traceable® Certificate.