AQ™ Brand Vials, MicroSolv
Supplier: MicroSolv Technology Corp
9512S-0CV 9512S-0CV-TD 9512S-0DCV 9532C-0CV 9532C-0DCV 9512C-0CV 9509S-WAV-AQ 9509S-WCV-AQ 9512C-0DCV 9512C-0CV-TD 9512C-0CV-T 9512S-0CV-T 9512S-0AV
97052-602PK 401.27 CAD
97052-602 97052-604 97052-606 97052-644 97052-648 97055-060 97055-448 97055-452 97055-456 97058-102 97035-136 97035-138 10040-096
AQ™ Brand Vials, MicroSolv
Vials
These vials are supplied without caps.
They are available in plastic or different types of glass materials, noticeably silanized/deactivated glass, RSA™ glass, and borosilicate glass.
Order Now
SPECIFICATIONS
Environmentally Preferable