Alligator clip wires
Supplier: United Scientific Supplies
WAG012-PK/4 WAG024-PK/6 WAG036-B WAG036-PK/6 WAG012-B WAG012-R WAG024-R WAG036-R WAG012-PK/6 WAG024-B
470219-731EA 16.25 CAD
470219-731 470219-720 470219-722 470219-733 470219-727 470219-729 470219-724 470219-735 470347-556 470219-737
Alligator clip wires
Wire
Alligator clip wires can connect to items that don't have banana plugs.
  • Protective Vinyl Sheath
  • Various Lengths and Pack Sizes Available
  • Flexible, 18-Gauge Stranded Wire Cord
A flexible, 18-gauge stranded wire cord has alligator clips at both ends.
Alligator clips have a protective vinyl sheath. They're available in various lengths and pack sizes.