Army Pattern Gouge, OR Grade, Sklar
Supplier: Sklar
40-6306 40-6312 40-6314 40-6318
10195-960EA 302.96 CAD
10195-960 10195-962 10195-964 10195-966
Army Pattern Gouge, OR Grade, Sklar
Gouges
Army pattern gouge is used to scoop bone and cartilage from curved bone surfaces.
  • Reusable, latex-free
  • Non sterile
One end of the gouge is curved or v-shaped and resembles a trough. A mallet may be used with the gouge if additional force is necessary.
Army Pattern Gouges are Stainless Steel and Industry Standard.