US Army Retractor, OR Grade, Sklar®
Supplier: Sklar
60-1085
10192-346PK 201.43 CAD
10192-346
US Army Retractor, OR Grade, Sklar®
Retractors
The US Army Retractor is used to expose surface layers of skin, commonly in plastic surgery procedures.
It is a double-ended retractor with a fenestrated handle
Censitrac Ready products are industry standard and constructed of stainless steel.
The blades at each end are angled 90° and also have a slightly curved, rounded lip. The blades also face in the same direction. US Army Retractors come in sets of 2.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.