Hank Uterine Dilators, OR Grade, Sklar®
Supplier: Sklar
90-4926 90-4918 90-4920 90-4925 90-4916 90-4914 90-4910 90-4912 90-4908
10194-064PK 1286.48 CAD
10194-064 10194-120 10194-122 10194-194 10194-262 10194-276 10194-296 10194-298 10199-270
Hank Uterine Dilators, OR Grade, Sklar®
Dilators
Sklar's® Hank Uterine Dilator is a manual, double-ended, probe-style OB/GYN instrument used for dilating the uterine cervix.
It is solid and non-flexible with smooth, blunt tips. Hank dilators are slightly curved and feature a stop ring at each end to prevent overinsertion. They are commonly used in procedures where gradual dilation of the cervix is necessary and are available in multiple sizes, graduated 5 mm each, to suit the needs of the patient. They are most frequently used in Dilation and Curretage and hysterectomy procedures.
Order Now
SPECIFICATIONS
Environmentally Preferable