Zebrafish Slides (Danio rerio)
Supplier: Avantor
470181-700EA 19.35 CAD
470181-700 470181-706 470181-708 470181-710
Zebrafish Slides (Danio rerio)
Slides Prepared Slides
Zebrafish are model organisms for vertebrate and genetic research.
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Zebrafish (Danio rerio) produce many offspring and have a short generation time for long term studies
We offer individual zebrafish slides of:
▪ Zebrafish: The entire fish is visible to the naked eye, but when viewed under a microscope, students can see the wide diversity of cell types found in one animal.
▪ Zebrafish, Hatchling : Typical hatchling, entire specimen.
▪ Zebrafish, Cleavage Stage: Illustrates cleavage stage.
▪ Zebrafish, Embryo: Entire embryo with yolk sac.
Embryos are great models to study vertebrate development because they are mostly transparent and developmental stages are easily visible.