Choice® SuperSorb Cleanroom-Laundered, Sealed-Edge Wiper, Berkshire
Supplier: Berkshire
CHSS12.12 CHSS09.14
89260-920CS 288.27 CAD
89260-920 76207-696 76670-464
Choice® SuperSorb Cleanroom-Laundered, Sealed-Edge Wiper, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Choice® SuperSorb is a two-ply, double knit, highly absorbent, ultrasonically sealed-edge, cleanroom-laundered wiper for reduced contamination. Made from 100% continuous filament knit polyester.
  • Critically low particles, fibers, ions, and extractables
  • High abrasion resistance
  • Chemically compatible with IPA, Acetone, and other solvents
  • Pinsonic design adds surface texture that aids in stubborn residue removal
Sealed edges increase durability and minimize fiber and particle release. Designed for the highest level of contamination control in critical processing applications, these wipers are excellent for the most critical spill control and cleaning applications.
Applications include sorbency for spill control in processing areas, stencil and print roll cleaning, cleaning of medical device products, applying and removing cleaning and disinfecting solutions. Steam autoclavable for aseptic environments.
Certifications: Complies with RoHS requirements.
Ordering information: Packaged in ISO class 4 (class 10) cleanroom.