Axygen® PCR Tube Strips, Corning
Supplier: Corning
Axygen®
PCR-0208-B
10011-762CS 4154.89 CAD
10011-762
Axygen® PCR Tube Strips, Corning
Tubes PCR Tubes
Axygen® 0.2 ml, thin-wall, 8-strip PCR tubes are available in an assortment of colors and are designed to fit a wide variety of thermal cyclers.
Ultra-thin and consistent wall thickness allows precise thermal transfer, even insertion cap ensures a tight fit to prevent evaporation.