Econo™ Sterile Nail Nipper, Floor Grade, Sklar
Supplier: Sklar
96-2666 96-2667
89023-988CS 1092.35 CAD
89023-988 89023-990
Econo™ Sterile Nail Nipper, Floor Grade, Sklar
Cutters
The Econo™ Sterile Nail Nipper is used for removing portions of the toenail or the nail plate.
This device features barrel spring, grooved, plier handles with a locking mechanism that is released upon squeezing the handles. This lock is intended to keep the sharp blades closed when the instrument is not being used.