Thermoelectric Effects Apparatus
Supplier: United Scientific Supplies
THEA01
470121-250EA 3100.4 CAD
470121-250
Thermoelectric Effects Apparatus
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Investigate the thermal characteristics of three electrical thermometry devices
Explore the features and behavior of a closed loop digital temperature control system
  • Investigate the thermal characteristics of three electrical thermometry devices
  • Explore the features and behavior of a closed loop digital temperature control system
The Thermoelectric Effects Apparatus consists of an oven containing a cooling fan, a massive metal block and electric heater supported on two thin struts, the three samples, and the control system sensor. A control unit provides the heating current and houses the digital temperature controller. The oven and the control unit are connected by three multi-pole cables.