Keyes Cutaneous Punch, OR Grade, Sklar
Supplier: Sklar
06-4144 06-4147 06-4146 06-4148 06-4145 06-4142 06-4143
10194-922EA 165.99 CAD
10194-922 10194-932 10194-924 10194-934 10194-926 10194-928 10194-930
Keyes Cutaneous Punch, OR Grade, Sklar
Biopsy Punches
Sklar's Keyes Cutaneous Punch is a dermal diagnostic instrument used to remove skin samples for biopsy.
The round sharp end is pushed into the skin and twisted slightly to cut away a small plug of skin. The solid knurled handle provides a gripping surface for the user who controls the depth of the excision. The Keyes punch is often used in gynecological biopsies where a deeper tissue sample is useful for the pathologist.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.