Econo™ Crile Hemostatic Forceps, Floor Grade, Sklar
Supplier: Sklar
21-447 21-452
89023-204PK 123.64 CAD
89023-204 89023-206
Econo™ Crile Hemostatic Forceps, Floor Grade, Sklar
Forceps
Econo™ Crile Hemostatic Forceps, also known as a Crile Hemostat, are used for clamping blood vessels or tissue before cauterization or ligation.
Horizontal serrations the entire length of the jaw, and jaws half the length of the shank make this instrument unique. There is a ratchet locking mechanism for a tight and secure hold on vessels. They may be used for soft tissue dissection no deeper than 6-8" or in laparotomy procedures.
Econo™ instruments are disposable.