Econo™ Hartmann Mosquito Forceps, Floor Grade, Sklar
Supplier: Sklar
21-409 21-421
89023-190PK 120.02 CAD
89023-190 89023-194
Econo™ Hartmann Mosquito Forceps, Floor Grade, Sklar
Forceps
Econo™ Hartmann Mosquito Forceps are designed as a hemostat and for small and fine tissue dissection in shallow wounds and procedures
Thanks to their fine tips, short and fully serrated jaws, these Mosquito Forceps can be used as a clamping instrument to halt blood flow, so that cauterization or ligation can be performed.
Hartmann Mosquito Forceps are shorter and heavier than Halstead Mosquito Forceps.
Econo™ instruments are disposable.
Originally for plastic and vascular surgeries, Hartmann Mosquito Forceps can be used in pediatric and ophthalmologic procedures.