Micro Surgery Needle Holder, OR Grade, Sklar
Supplier: Sklar
98-3180 98-3186 98-3188 98-3182 98-3184 98-3185 98-3190 98-3192 98-7820 98-3183
10185-310EA 1252.69 CAD
10185-310 10185-312 10192-232 10194-620 10194-622 10194-624 10194-626 10194-628 75818-100 75995-892
Micro Surgery Needle Holder, OR Grade, Sklar
Forceps
Sklar's® micro needle holder is most commonly used to hold fine suture needles in very small sites or microsurgical procedures.
  • Ergonomical handles are designed with texturized grips for comfort
  • Available in various lengths
Micro needle holder is available in various curvature, straight or curved, with or without a catch.
Order Now
SPECIFICATIONS
Environmentally Preferable