EcoClean® 60 Cotton Nonwoven Cleanroom Wiper, Berkshire
Supplier: Berkshire
ECW60.090920 ECW60.121220
89097-480CS 1175.79 CAD
89097-480 89097-482
EcoClean® 60 Cotton Nonwoven Cleanroom Wiper, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
EcoClean® products are nonwoven materials 100% biodegradable and sustainable products. It is composed of U.S.-sourced cotton fibers and is manufactured by a proprietary process for low fiber release and high strength. Unlike the traditional cotton twill, it has low ionic traces and low fibers.
  • Heat resistant
  • Highly absorbent and durable
  • Extremely soft
  • Resists build-up of static charge
It is in flat sheet format for cleaning high temperature production equipment.
Low linting work surface material. Removing aqueous and solvent spills. Wiping of optics, other sensitive surfaces and good upgrade from gauze.
Certifications: Certified for contact with food per EC Regulation 1935(2004) - FDA 21 CFR - Food contact compliant. For use in ISO class 5 to 8 (class 100 to 100,000) cleanroom applications.
Ordering information: 20 packs per case (double packed).