Wire Shelving Systems with Conductive Bins, Quantum Storage Systems
Supplier: QUANTUM STORAGE SYSTEMS
WR12-101CO WR12-104CO WR12-108CO WR12-110CO WR12-105CO WR12-106CO WR12-114CO WR12-116CO WR12-102CO WR12-107CO WR12-109CO WR12-103CO WR6-36-1236-101CO WR6-36-1236-102CO WR6-36-1236-107CO WR6-36-1236-109CO
10805-440EA 5253.17 CAD
10805-440 10805-442 10805-444 10805-446 10805-448 10805-450 10805-830 10805-832 10805-836 10805-838 10805-840 10805-842 10809-728 10809-730 10809-732 10809-734
Wire Shelving Systems with Conductive Bins, Quantum Storage Systems
Furniture Shelves
These shelve units come standard with conductive bins, conductive split sleeves and a drag chain to be attached to the bottom shelf helping to ground the shelving unit.
  • Eliminate the build up of a conductive charge
  • Protect the electronic components being stored
ESD-safe.
Delivery information: Supplied with black bins.