Surgi-OR™ Hartmann Mosquito Forceps, Mid Grade, Sklar
Supplier: Sklar
95-409 95-421 95-431
89023-610EA 61.81 CAD
89023-610 89023-612 89023-616
Surgi-OR™ Hartmann Mosquito Forceps, Mid Grade, Sklar
Forceps
Surgi-OR™ Hartmann Mosquito Forceps are used as both a hemostat and for fine tissue dissection in shallow wounds and procedures
Known for their fine tips, short, and fully serrated jaws, these Mosquito Forceps can be used as a clamping instrument to halt blood flow so that cauterization or ligation can be performed. They are also used for holding small, fine sutures. Hartmann Mosquito Forceps are shorter and heavier than Halstead Mosquito Forceps.
Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from high quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers.
Mostly used in plastic and vascular surgeries, Hartmann Mosquito Forceps can also be used in pediatric and ophthalmologic procedures.