Econo™ Iris Dressing Forceps, Floor Grade, Sklar
Supplier: Sklar
21-123 21-124 21-135 21-146 21-151 21-128 21-129TP 21-142
89023-038PK 86.01 CAD
89023-038 89023-040 89023-044 89023-046 89023-050 89023-052 76192-370 76192-372 76192-374
Econo™ Iris Dressing Forceps, Floor Grade, Sklar
Forceps
Econo™ Iris Dressing Forceps are thumb style forceps used for placing or removing dressings, although they were designed to hold materials such as gauze
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Econo™ Iris Dressing Forceps are thumb style forceps commonly used for placing or removing dressings in many types of procedures. Although Dressing Forceps were originally designed to specifically hold materials such as gauze, they have come to have a variety of additional functions. They may be used for removing foreign objects or dead tissue from wounds, as well as other small surgical materials such as tubing. Serrated jaws assist in a having a secure hold on items. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.