VWR® Built In Undercounter Refrigerators with Natural Refrigerants
Supplier: VWR International
HCUCBI0404 HCUCBI0404G HCUCBI0404AD HCUCBI0404SS HCUCBI0404LH HCUCBI0404GLH HCUCBI0404ADLH HCUCBI0404SSLH
10819-880EA 3132.57 CAD
10819-880 10819-882 10819-884 10819-886 76210-106 76210-108 76210-110 76210-112
VWR® Built In Undercounter Refrigerators with Natural Refrigerants
Refrigerators
These undercounter laboratory refrigerators provide lab quality cold storage in a space-saving design.
  • Integrated temperature alarms
  • Keyed door locks
  • Forced draft circulation
  • Precise microprocessor temperature control
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
Built-in units are designed with front-facing ventilation systems, allowing easy installation into a bench or countertop. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Certifications: *Select models ENERGY STAR certified: 10819-880, 10819-884, 76210-110, 10819-882, 76210-108, 76210-106, 76210-112, 10819-886
Ordering information: All units have a two-year parts and labor warranty. Select models come with an additional three-year compressor parts warranty. (Please refer to unit datasheets to confirm).