Manganese(IV) oxide tech. 90%, Technical Grade, activated

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Manganese dioxide, Manganese(IV) oxide, activated, Manganese peroxide, Mangan(IV)-oxid

L13253-09 L13253-36
AAL13253-09EA 49.45 CAD
AAL13253-09 AAL13253-36
Manganese(IV) oxide tech. 90%, Technical Grade, activated
Manganese (IV) oxide

Manganese(IV) oxide is suitable for use in batteries. It is widely used for the selective oxidation of allylic and benzylic alcohols to aldehydes or ketones, as a co-oxidant in combination with 1,4-benzoquinone and in the allylic oxidation of olefins catalyzed by Palladium(II)­ acetate. It acts a reagent for oxidations. It is the source of manganese and all its compounds, largely used in manufacturing of manganese steel, for making amethyst glass, decolorizing glass, painting on porcelain, faience and majolica. The precipitate is used in electrotechnics, pigments, browning gun barrels, drier for paints and varnishes, printing and dyeing textiles.


Purity: 90% CAS No.: 1313-13-9 Molecular Formula: MnO2 Formula Weight: 86.94 Melting Point: ca 535° dec. Merck Reference: 14,5730 MDL No.: MFCD00003463

Formula: MnO₂
MW: 86.94 g/mol
Melting Pt: 535 (dec.)
Density: 5.026
Storage Temperature: Ambient
MDL Number: MFCD00003463
CAS Number: 1313-13-9
EINECS: 215-202-6
Merck Index: 14,05730

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR