4-Iodo-2-methoxynicotinaldehyde 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66984-03
H66984-MD
AAH66984-03EA
282.82
CAD
AAH66984-03
AAH66984-MD
4-Iodo-2-methoxynicotinaldehyde 95%
4-Iodo-2-methoxynicotinaldehyde
Purity: 95%
CAS No.: 158669-26-2
Molecular Formula: C7H6INO2
Formula Weight: 263.03
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Sensitivity: Light Sensitive
Beilstein Reference: 7020882
MDL No.: MFCD05662396
Formula:
C₇H₆INO₂ Storage Temperature: Freezer |
MDL Number:
MFCD05662396 CAS Number: 158669-26-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...