3,5-Dichloropyridine-2-carboxamide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66981-03
H66981-MD
AAH66981-03EA
565.64
CAD
AAH66981-03
AAH66981-MD
3,5-Dichloropyridine-2-carboxamide 97%
3,5-Dichloropyridine-2-carboxamide
Purity: 97%
CAS No.: 5468-71-3
Molecular Formula: C6H4Cl2N2O
Formula Weight: 191.01
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Beilstein Reference: 136067
MDL No.: MFCD20529142
Formula:
C₆H₄Cl₂N₂O Storage Temperature: Freezer |
MDL Number:
MFCD20529142 CAS Number: 5468-71-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...