4-[(4-Aminophenyl)azo]-2-chlorophenol 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66952-03
H66952-MD
AAH66952-03EA
496.12
CAD
AAH66952-03
AAH66952-MD
4-[(4-Aminophenyl)azo]-2-chlorophenol 95%
4-[(4-Aminophenyl)azo]-2-chlorophenol
Purity: 95%
CAS No.: 669092-47-1
Molecular Formula: C12H10ClN3O
Formula Weight: 247.68
Physical Form: Solid
Formula:
C₁₂H₁₀ClN₃O Storage Temperature: Ambient |
MDL Number:
MFCD26401540 CAS Number: 669092-47-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...