5-Carboxyfluorescein single isomer 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Spiro(isobenzofuran-1(3H),9'-(9H)xanthene)-5-carboxylic acid, 3',6'-dihydroxy-3-oxo-, 5-FAM, 3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-5-carboxylic acid, 5-Carboxyfluorescein hydrate
H66949-03
H66949-MD
AAH66949-03EA
788.42
CAD
AAH66949-03
AAH66949-MD
5-Carboxyfluorescein single isomer 97%
5-Carboxyfluorescein
Purity: 97%
CAS No.: 76823-03-5
Molecular Formula: C21H12O7
Formula Weight: 376.32
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Beilstein Reference: 57037
MDL No.: MFCD00036874
Formula:
C₂₁H₁₂O₇.xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00036874 CAS Number: 76823-03-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...