Dihydrofuran-3(2H)-one 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Oxolan-3-one, Tetrahydrofuran-3-one

H66940-03 H66940-06
AAH66940-03EA 70.63 CAD
AAH66940-03 AAH66940-06
Dihydrofuran-3(2H)-one 95%
Dihydrofuran-3(2H)-one

Purity: 95% CAS No.: 22929-52-8 Molecular Formula: C4H6O2 Formula Weight: 86.09 Storage Temperature: 2°C to 8°C Physical Form: Liquid Beilstein Reference: 105200 MDL No.: MFCD07778393

Formula: C₄H₆O₂
MW: 86.09 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07778393
CAS Number: 22929-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR