5-Formyl-2-picoline 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66926-03
H66926-06
H66926-MD
AAH66926-03EA
132.88
CAD
AAH66926-03
AAH66926-06
AAH66926-MD
5-Formyl-2-picoline 96%
5-Formyl-2-picoline
Purity: 96%
CAS No.: 53014-84-9
Molecular Formula: C7H7NO
Formula Weight: 121.14
Flash Point: 87°(189°F)
Physical Form: Liquid
Density: 1.084
Refractive Index: 1.5460
Sensitivity: Moisture Sensitive
Boiling Point: 95°/20mm
Beilstein Reference: 107496
MDL No.: MFCD08272279
Formula:
C₇H₇NO Boiling Pt: 95 °C/20 mm Density: 1.084 Flash Pt: 87 °C(189 °F) Storage Temperature: Ambient |
MDL Number:
MFCD08272279 CAS Number: 53014-84-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...